Stampa…

Anti-TNIK Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: United States Biological
USBIT8172-05E-APCEA 1200 CHF
USBIT8172-05E-APC
Anti-TNIK Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000
Do not freeze!

Type: Primary
Antigen: TNIK (TRAF2 and NCK interacting kinase)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR