Stampa…
Anti-TNIK Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8172-05DEA 825 CHF
USBIT8172-05D
Anti-TNIK Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: TNIK (TRAF2 and NCK interacting kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR