Stampa…
Anti-TNIK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornitore: United States Biological
USBIT8172-05C-HRPEA 1200 CHF
USBIT8172-05C-HRP
Anti-TNIK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: TNIK (TRAF2 and NCK interacting kinase)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Mehr About VWR