Stampa…

Anti-TNIK Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT8172-05C-BIOTEA 1200 CHF
USBIT8172-05C-BIOT
Anti-TNIK Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TNIK (TRAF2 and NCK interacting kinase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR