Stampa…

Anti-TRAF2 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8171-94NEA 774 CHF
USBIT8171-94N
Anti-TRAF2 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:128000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR