Stampa…
Anti-TRAF2 Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBIT8171-94L-BIOTEA 1200 CHF
USBIT8171-94L-BIOT
Anti-TRAF2 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR