Stampa…

Anti-TRAF2 Mouse Monoclonal Antibody [clone: 9F648]

Fornitore: United States Biological
USBIT8171-94KEA 753 CHF
USBIT8171-94K
Anti-TRAF2 Mouse Monoclonal Antibody [clone: 9F648]
Anticorpi
Recommended dilution: ELISA: 1:2000

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: monoclonal
Clone: 9F648
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR