Stampa…

Anti-TRAF Interacting Protein TANK Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8171-76EA 889 CHF
USBIT8171-76
Anti-TRAF Interacting Protein TANK Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TRAF Interacting Protein TANK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR