Stampa…
Anti-TAAR1 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8170-01C2EA 1150 CHF
USBIT8170-01C2
Anti-TAAR1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 25-35 ?g/ml
Type: Primary
Antigen: TAAR1 (trace amine associated receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR