Stampa…

Anti-TRA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9H185]

Fornitore: United States Biological
USBIT8115AEA 930 CHF
USBIT8115A
Anti-TRA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9H185]
Anticorpi
Type: Primary
Antigen: TRA (T cell receptor alpha locus)
Clonality: monoclonal
Clone: 9H185
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR