Stampa…

Anti-TRA1-81 Mouse Monoclonal Antibody [clone: 5C3]

Fornitore: United States Biological
USBIT8115-10EA 962 CHF
USBIT8115-10
Anti-TRA1-81 Mouse Monoclonal Antibody [clone: 5C3]
Anticorpi
Recommended dilution: Flow Cytometry: 10-20 ?g/ml

Type: Primary
Antigen: TRA1-81
Clonality: monoclonal
Clone: 5C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR