Stampa…

Anti-NR2C1 Rat Monoclonal Antibody [clone: 6D805]

Fornitore: United States Biological
USBIT8105-05EA 847 CHF
USBIT8105-05
Anti-NR2C1 Rat Monoclonal Antibody [clone: 6D805]
Anticorpi
Recommended dilution: Flow Cytometry: 1:1000; Immunoprecipitation: 1:200

Type: Primary
Antigen: NR2C1 (nuclear receptor subfamily 2, group C, member 1)
Clonality: monoclonal
Clone: 6D805
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR