Stampa…

Anti-Toxoplasma gondii Mouse Monoclonal Antibody [clone: 10D29]

Fornitore: United States Biological
USBIT8076-10FEA 641 CHF
USBIT8076-10F
Anti-Toxoplasma gondii Mouse Monoclonal Antibody [clone: 10D29]
Anticorpi
Recommended dilution: Immunofluorescence: 1:10-1:50; ELISA: 1:20-1:200; Western Blot: 1:20-1:200

Type: Primary
Antigen: Toxoplasma gondii
Clonality: monoclonal
Clone: 10D29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR