Stampa…
Anti-Toxoplasma gondii Goat Polyclonal Antibody
Fornitore: United States Biological
USBIT8075-02EA 589 CHF
USBIT8075-02
Anti-Toxoplasma gondii Goat Polyclonal Antibody
Anticorpi
Recommended dilution: Immunofluorescence: 1:1000
Type: Primary
Antigen: Toxoplasma gondii
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Visualizza i codici
Learn more
About VWR
Mehr About VWR