Stampa…

Anti-CRTC2 Rabbit polyclonal antibody

Fornitore: United States Biological
USBIT8067-21BEA 632 CHF
USBIT8067-21B
Anti-CRTC2 Rabbit polyclonal antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: CRTC2 (CREB regulated transcription coactivator 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR