Stampa…

Anti-Topoisomerase Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8066-01EA 725 CHF
USBIT8066-01
Anti-Topoisomerase Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunoprecipitation: 0.5-2.5 ?g/mg; Western Blot: 1:1000-1:10000

Type: Primary
Antigen: Topoisomerase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR