Stampa…
Anti-TOP2B Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8065-04AEA 784 CHF
USBIT8065-04A
Anti-TOP2B Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: TOP2B (topoisomerase (DNA) II beta 180kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Mehr About VWR