Stampa…

Anti-TOP1 Mouse Monoclonal Antibody [clone: 11C887]

Fornitore: United States Biological
USBIT8064-99DEA 733 CHF
USBIT8064-99D
Anti-TOP1 Mouse Monoclonal Antibody [clone: 11C887]
Anticorpi
Type: Primary
Antigen: TOP1 (topoisomerase (DNA) I)
Clonality: monoclonal
Clone: 11C887
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR