Stampa…

Anti-TOM70 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8064-18EA 756 CHF
USBIT8064-18
Anti-TOM70 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TOM70
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR