Stampa…
Anti-TOLLIP Rat Monoclonal Antibody [clone: 6D801]
Fornitore: United States Biological
USBIT8063-50EA 1060 CHF
USBIT8063-50
Anti-TOLLIP Rat Monoclonal Antibody [clone: 6D801]
Anticorpi
Recommended dilution: Western Blot: 5 ?g/ml
Type: Primary
Antigen: TOLLIP (toll interacting protein)
Clonality: monoclonal
Clone: 6D801
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Mehr About VWR