Stampa…

Anti-Toll Like Receptor 12 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIT8051-22AEA 747 CHF
USBIT8051-22A
Anti-Toll Like Receptor 12 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: Toll Like Receptor 12
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR