Stampa…

Anti-TLR9 Mouse Monoclonal Antibody [clone: 6A643]

Fornitore: United States Biological
USBIT8050-93BEA 813 CHF
USBIT8050-93B
Anti-TLR9 Mouse Monoclonal Antibody [clone: 6A643]
Anticorpi
Recommended dilution: Immunohistology: 1:10; Flow Cytometry: 1:10; Western Blot: 1:10

Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: monoclonal
Clone: 6A643
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR