Stampa…
Anti-TLR9 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8050-91JEA 739 CHF
USBIT8050-91J
Anti-TLR9 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2?g/ml
Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Mehr About VWR