Stampa…

Anti-TLR9 Mouse Monoclonal Antibody (Biotin) [clone: 5G5]

Fornitore: United States Biological
USBIT8050-90P1EA 978 CHF
USBIT8050-90P1
Anti-TLR9 Mouse Monoclonal Antibody (Biotin) [clone: 5G5]
Anticorpi
Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: monoclonal
Clone: 5G5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR