Stampa…

Anti-TLR9 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9G286]

Fornitore: United States Biological
USBIT8050-90F1EA 966 CHF
USBIT8050-90F1
Anti-TLR9 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9G286]
Anticorpi
Recommended dilution: Flow Cytometry: 1:50; Western Blot: 1:50; Immunochemistry: 1:50

Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: monoclonal
Clone: 9G286
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR