Stampa…

Anti-TLR9 Mouse Monoclonal Antibody [clone: 6D84]

Fornitore: United States Biological
USBIT8050-90EEA 1020 CHF
USBIT8050-90E
Anti-TLR9 Mouse Monoclonal Antibody [clone: 6D84]
Anticorpi
Recommended dilution: Immunochemistry: 10-20 ?g/ml; Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: monoclonal
Clone: 6D84
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR