Stampa…

Anti-TLR8 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8050-85NEA 632 CHF
USBIT8050-85N
Anti-TLR8 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: TLR8 (toll-like receptor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR