Stampa…

Anti-TLR8 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIT8050-85H-FITCEA 1200 CHF
USBIT8050-85H-FITC
Anti-TLR8 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: TLR8 (toll-like receptor 8)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR