Stampa…

Anti-TLR8 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8050-85HEA 825 CHF
USBIT8050-85H
Anti-TLR8 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100; Western Blot: 1:100-1:500

Type: Primary
Antigen: TLR8 (toll-like receptor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR