Stampa…
Anti-TLR8 Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBIT8050-85F-BIOTEA 1200 CHF
USBIT8050-85F-BIOT
Anti-TLR8 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TLR8 (toll-like receptor 8)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR