Stampa…

Anti-TLR8 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8050-83BEA 889 CHF
USBIT8050-83B
Anti-TLR8 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TLR8 (toll-like receptor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR