Stampa…

Anti-TLR7 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8050-73AEA 660 CHF
USBIT8050-73A
Anti-TLR7 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR