Stampa…

Anti-TLR7 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8050-72LEA 889 CHF
USBIT8050-72L
Anti-TLR7 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-3?g/ml Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR