Stampa…

Anti-TLR7 Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT8050-72G-BIOTEA 1200 CHF
USBIT8050-72G-BIOT
Anti-TLR7 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR