Stampa…

Anti-TLR7 Mouse Monoclonal Antibody [clone: 10F536]

Fornitore: United States Biological
USBIT8050-71BEA 913 CHF
USBIT8050-71B
Anti-TLR7 Mouse Monoclonal Antibody [clone: 10F536]
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: monoclonal
Clone: 10F536
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR