Stampa…

Anti-TLR6 Mouse Monoclonal Antibody (Biotin) [clone: 11E375]

Fornitore: United States Biological
USBIT8050-53NEA 899 CHF
USBIT8050-53N
Anti-TLR6 Mouse Monoclonal Antibody (Biotin) [clone: 11E375]
Anticorpi
Type: Primary
Antigen: TLR6 (toll-like receptor 6)
Clonality: monoclonal
Clone: 11E375
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR