Stampa…

Anti-TLR5 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11E373]

Fornitore: United States Biological
USBIT8050-41AEA 878 CHF
USBIT8050-41A
Anti-TLR5 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11E373]
Anticorpi
Type: Primary
Antigen: TLR5 (toll-like receptor 5)
Clonality: monoclonal
Clone: 11E373
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR