Stampa…

Anti-TLR4/MD-2 Complex Rat Monoclonal Antibody [clone: 6D82]

Fornitore: United States Biological
USBIT8050-35BEA 687 CHF
USBIT8050-35B
Anti-TLR4/MD-2 Complex Rat Monoclonal Antibody [clone: 6D82]
Anticorpi
Type: Primary
Antigen: TLR4/MD-2 Complex
Clonality: monoclonal
Clone: 6D82
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR