Stampa…

Anti-TLR4 Mouse Monoclonal Antibody [clone: 3B6]

Fornitore: United States Biological
USBIT8050-34PEA 988 CHF
USBIT8050-34P
Anti-TLR4 Mouse Monoclonal Antibody [clone: 3B6]
Anticorpi
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: monoclonal
Clone: 3B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR