Stampa…

Anti-TLR4 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 11E372]

Fornitore: United States Biological
USBIT8050-34NEA 971 CHF
USBIT8050-34N
Anti-TLR4 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 11E372]
Anticorpi
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: monoclonal
Clone: 11E372
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR