Stampa…

Anti-TLR4 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8050-33EA 756 CHF
USBIT8050-33
Anti-TLR4 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR