Stampa…

Anti-TLR3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11n194]

Fornitore: United States Biological
USBIT8050-24CEA 833 CHF
USBIT8050-24C
Anti-TLR3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11n194]
Anticorpi
Do not freeze!

Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: monoclonal
Clone: 11n194
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR