Stampa…

Anti-TLR3 Mouse Monoclonal Antibody [clone: 9H184]

Fornitore: United States Biological
USBIT8050-20LEA 1050 CHF
USBIT8050-20L
Anti-TLR3 Mouse Monoclonal Antibody [clone: 9H184]
Anticorpi
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: monoclonal
Clone: 9H184
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR