Stampa…

Anti-TLR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13A75]

Fornitore: United States Biological
USBIT8050-17D2EA 938 CHF
USBIT8050-17D2
Anti-TLR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13A75]
Anticorpi
Do not freeze!

Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: 13A75
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR