Stampa…

Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: 6A629]

Fornitore: United States Biological
USBIT8050-15DEA 711 CHF
USBIT8050-15D
Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: 6A629]
Anticorpi
Recommended dilution: Immunohistology: 1:10; Flow Cytometry: 1:10; Western Blot: 1:10

Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: 6A629
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR