Stampa…

Anti-TLR2 Mouse Monoclonal Antibody [clone: T2.5]

Fornitore: United States Biological
USBIT8050-11JEA 1130 CHF
USBIT8050-11J
Anti-TLR2 Mouse Monoclonal Antibody [clone: T2.5]
Anticorpi
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: T2.5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR