Stampa…

Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: 10B1261]

Fornitore: United States Biological
USBIT8050-11D4EA 725 CHF
USBIT8050-11D4
Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: 10B1261]
Anticorpi
Recommended dilution: Flow Cytometry:1:10;

Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: 10B1261
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR