Stampa…

Anti-TLR2 Mouse Monoclonal Antibody [clone: 9G275]

Fornitore: United States Biological
USBIT8050-09SEA 792 CHF
USBIT8050-09S
Anti-TLR2 Mouse Monoclonal Antibody [clone: 9G275]
Anticorpi
Recommended dilution: Immunofluorescence: 1:50; Flow Cytometry: 1:50; Immunoprecipitation: 1:50

Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: 9G275
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR