Stampa…

Anti-TLR2 Mouse Monoclonal Antibody [clone: 6D78]

Fornitore: United States Biological
USBIT8050-09GEA 653 CHF
USBIT8050-09G
Anti-TLR2 Mouse Monoclonal Antibody [clone: 6D78]
Anticorpi
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: 6D78
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR