Stampa…

Anti-TLR1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8050-01NEA 739 CHF
USBIT8050-01N
Anti-TLR1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-2?g’ml

Type: Primary
Antigen: TLR1 (toll-like receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR